Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSK Rabbit anti-Human, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Manufacturer:  LIFE TECHNOLOGIES 703673

Encompass

Catalog No. 70-367-3


Add to cart

Description

Description

This antibody is predicted to react with Monkey, Mouse, Pig. Recombit rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombit rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

C-terminal Src kinase (CSK) is a non-receptor protein tyrosine kinase that plays an important role in the regulation of cell growth, differentiation, migration, and immune responses. CSK phosphorylates tyrosine residues located in the C-terminal tails of Src-family kinases including LCK, SRC, HCK, FYN, LYN, CSK, and YES1. CSK is expressed ubiquitously as a protein of approximately 50 KDa in the cytosol, but a fraction distributes to lipid rafts due to interaction with the lipid raft-associated protein PAG (Csk-binding protein).
Specifications

Specifications

CSK
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P41240
CSK
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Human
Western Blot
5H3L21
Unconjugated
CSK
Liquid
CSK
Protein corresponding to human CSK [aa73-aa178]
Protein A
RUO
Antibody
1445
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit