Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
csl/RBPJK Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31089425UL
Description
csl/RBPJK Polyclonal specifically detects csl/RBPJK in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
csl/RBPJK | |
Polyclonal | |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunoprecipitation | |
CBF-1, csl, H-2K binding factor-2, IGKJRB1RBPSUHCBF1, IGKJRBRBP-J kappa, immunoglobulin kappa J region recombination signal binding protein 1, J kappa-recombination signal-binding protein, KBF2, RBP-JK, RBPJKMGC61669, RBP-Jrecombining binding protein suppressor of hairless (Drosophila), recombination signal binding protein for immunoglobulin kappa J region, recombining binding protein suppressor of hairless, Renal carcinoma antigen NY-REN-30, SUH, suppressor of hairless homolog | |
The immunogen is a synthetic peptide directed towards the C terminal region of human csl/RBPJK (NP_976028). Peptide sequence RPHCSAAGAILRANSSQVPPNESNTNSEGSYTNASTNSTSVTSSTATVVS | |
25 μg | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
3516 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction