Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSMD1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA519235
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
CSMD1 CUB and Sushi multiple domains 1 is a protein that in humans is encoded by the CSMD1gene. It is a potential tumour suppressor, the deletion of which may result in head and neck carcinomas behaving more aggressively. CSMD1 is part of the complement system that defends against pathogens through either the classical pathway or the alternative pathway. Located primarily in nerve growth cones, CSMD1 blocks the classical pathway of the immune system and is thought to be involved in tumor suppression, as defects in the gene encoding CSMD1 are associated with squamous cell carcinomas.
Specifications
CSMD1 | |
Polyclonal | |
Unconjugated | |
CSMD1 | |
B930082H09; Csmd1; CUB and sushi domain-containing protein 1; CUB and Sushi multiple domains 1; CUB and sushi multiple domains protein 1; KIAA1890; Mdcr1; mKIAA1890; multiple domain complement regulator 1; PPP1R24; protein phosphatase 1, regulatory subunit 24; UNQ5952/PRO19863 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
64478 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96PZ7 | |
CSMD1 | |
Synthetic peptide sequence (KGVREVNETVTKTP) corresponding to the internal amino acids of CSMD1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction