Learn More
CSNK1G2 Mouse anti-Human, Clone: OTI4H3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802852
Catalog No. 50-166-6200
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling.
Specifications
Specifications
CSNK1G2
Monoclonal
Unconjugated
CSNK1G2
Lyophilized
CSNK1G2
Human recombit protein fragment corresponding to amino acids 163-415 of human CSNK1G2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI4H3
PBS with 8% trehalose and no preservative; pH 7.3
P78368
CK1g2
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1455