Learn More
Abnova™ CSNK2A2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00001459T02
Catalog No. 89-940-779
Add to cart
Specifications
Specifications
Western Blot
293T
NM_001896.2
pCMV-CSNK2A2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
CSNK2A2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 41.2kDa
Transient overexpression cell lysate was tested with anti-CSNK2A2 antibody (Mfr. No. H00001459-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)