Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSP alpha Rabbit anti-Guinea Pig, Human, Mouse, Ovine, Rabbit, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549570

Catalog No. PA549570


Add to cart

Description

Description

May have an important role in presynaptic function. May be involved in calcium-dependent neurotransmitter release at nerve endings.
Specifications

Specifications

CSP alpha
Polyclonal
TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH 7.4
Q9H3Z4, P60904, P60905
DnaJ homolog subfamily C member 5, Cysteine string protein, CSP, DNAJC5, CSP
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100352309, 100728482, 101108591, 13002, 79130, 80331
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DNAJC5
Liquid
DNAJC5
synthetic peptide corresponding to 1-100aa of human CSP
Protein A
RUO
Antibody
Polyclonal
Guinea Pig, Human, Mouse, Ovine, Rabbit, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit