Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CST3 Goat anti-Human, Polyclonal , Abnova™

Goat polyclonal antibody raised against native human CST3.

Manufacturer:  Abnova Corporation PAB29782

Catalog No. 89-934-013


Add to cart

Description

Description

The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions, where they appear to provide protective functions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes the most abundant extracellular inhibitor of cysteine proteases, which is found in high concentrations in biological fluids and is expressed in virtually all organs of the body. A mutation in this gene has been associated with amyloid angiopathy. Expression of this protein in vascular wall smooth muscle cells is severely reduced in both atherosclerotic and aneurysmal aortic lesions, establishing its role in vascular disease. [provided by RefSeq]

Specifications

Specifications

CST3
Unconjugated
ELISA Western Blot The optimal working dilution should be determined by the end user.
P01034
ARMD11/MGC117328
Goat
IgG
1 mg
Store at 4°C.
Polyclonal
Human
ELISA, Western Blot
Goat polyclonal antibody raised against native human CST3.
In PBS, pH 7.2 (0.09% sodium azide)
Liquid
CST3
Native purified human CST3.
Antigen Affinity Purified
RUO
Primary
1471
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit