Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CST4 Mouse anti-Human, Clone: OTI2H10, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504135S

Catalog No. 50-167-4885


Add to cart

Description

Description

The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes a type 2 salivary cysteine peptidase inhibitor. The protein is an S-type cystatin, based on its high level of expression in saliva, tears and seminal plasma. The specific role in these fluids is unclear but antibacterial and antiviral activity is present, consistent with a protective function.
Specifications

Specifications

CST4
Monoclonal
0.89 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P01036
cystatin S
Mouse
IgG2b
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1472
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2H10
Unconjugated
CST4
Liquid
CST4
Full length human recombit protein of human CST4 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit