Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSTF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120963

This item is not returnable.
View return policy
Description
Positive test controls include: U2OS, MCF7, SW480, Mouse thymus, Rat thymus. The target is usually found in the following locations: Nucleus.
CSTF1 Polyclonal antibody specifically detects CSTF1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
CSTF1 | |
Polyclonal | |
Unconjugated | |
CSTF1 | |
1700057K18Rik; AI788832; CF-1 50 kDa subunit; cleavage stimulation factor 50 kDa subunit; Cleavage stimulation factor subunit 1; cleavage stimulation factor, 3' pre-RNA, subunit 1; cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kD; cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa; CSTF 50 kDa subunit; Cstf1; CstF50; CstF-50; CstFp50 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human CSTF1 (NP_001315.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
3.73 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q05048, Q5BJQ6, Q99LC2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1477, 311670, 67337 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title