Learn More
CSTF1 Rabbit anti-Human, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 155371AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0295
Add to Cart
Description
Description
This gene encodes one of three subunits which combine to form cleavage stimulation factor (CSTF). CSTF is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. Similar to mammalian G protein beta subunits, this protein contains transducin-like repeats. Several transcript variants with different 5′ UTR, but encoding the same protein, have been found for this gene.
Specifications
Specifications
CSTF1
Polyclonal
Unconjugated
CSTF1
CSTF1
CSTF1 Fusion Protein Ag7874
150 μL
Primary
Human
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q05048
Rabbit
Antigen Affinity Chromatography
RUO
1477
-20°C
Liquid
Product Suggestions
Product Suggestions