Learn More
Abnova™ CSTF2T 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00023283T01
Catalog No. 89-940-835
Add to cart
Specifications
Specifications
Western Blot
293T
NM_015235
pCMV-CSTF2T full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
CSTF2T 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 64.4kDa
Transient overexpression cell lysate was tested with anti-CSTF2T antibody (Mfr. No. H00023283-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)