Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSTF2T Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB29882
Description
Rabbit polyclonal antibody raised against partial synthetic protein of human CSTF2T.
- High specificity
- High reactivity
Specifications
CSTF2T | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial synthetic protein of human CSTF2T. | |
In PBS, pH 7.4 (2% sucrose, 0.09% sodium azide). | |
CstF-64T/DKFZp434C1013/KIAA0689 | |
Rabbit | |
Affinity purification | |
RUO | |
23283 | |
Store at 4°C for up to 1 week. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunofluorescence, Western Blot | |
Unconjugated | |
Immunofluorescence (1:250) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Q9H0L4 | |
CSTF2T | |
A synthetic peptide corresponding to amino acids 460-509 of human CSTF2T. | |
100 μL | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction