Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CT45A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548334
Description
CT45A4 Polyclonal antibody specifically detects CT45A4 in Human samples. It is validated for Western Blot.Specifications
CT45A4 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8NHU0 | |
CT45A3 | |
KLH conjugated synthetic peptide between 106-135 amino acids from the Central region of human CT45A4/CT45A3/CT45A2/CT45A1/CT45A6. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CT45A3 | |
cancer/testis antigen 45-3; Cancer/testis antigen 45-4; cancer/testis antigen 45A3; Cancer/testis antigen 45A4; cancer/testis antigen CT45-3; cancer/testis antigen family 45 member A3; Cancer/testis antigen family 45 member A4; cancer/testis antigen family 45, member A3; cancer/testis antigen family 45, member A4; CT45.3; CT45.4; CT45-3; CT45-4; CT45A3; CT45A4 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
441519 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction