Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CT55 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA5139845

Encompass

Catalog No. PIPA5139845


Add to Cart
This item is not returnable. View return policy

Description

Description

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GNVPLKVGQK VNVVVEEDKP HYGLRAIKVD VVPRHLYGAG PSDSGTRVLI G

CT55 Polyclonal antibody specifically detects CT55 in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CT55
Polyclonal
Unconjugated
CT55
BJHCC20A; BRCA2-interacting protein; cancer/testis antigen 55; CT55; CXorf48; tumor antigen BJ-HCC-20
Rabbit
Antigen affinity chromatography
RUO
54967
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 μg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8WUE5
CT55
Recombinant protein corresponding to Human CT55
100 μL
Primary
Human
Antibody
IgG
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit