Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CTA 056 Available on GSA/VA Contract for Federal Government customers only.

ITK inhibitor

Manufacturer:  Tocris Bioscience™ 4726/10

Catalog No. 472610


Add to cart
This item is not returnable. View return policy

Description

Description

Interleukin-2-inducible T-cell kinase (ITK) inhibitor (IC50 values are 0.1, 0.4 and 5 μM for ITK, BTK and ETK, respectively). Inhibits growth of cancer cells expressing ITK, including MOLT-4 and Jurkat cells in vitro. Induces apoptosis in Jurkat cells. Inhibits MOLT-4 xenograft tumor growth in mice.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

1,5-Dihydro-7-(phenylmethyl)-1-[3-(1-piperidinyl)propyl]-2-[4-(4-pyridinyl)phenyl]-6H-imidazo[4,5-g]quinoxalin-6-one
1265822-30-7
554.68
Target: Other Kinase Inhibitors
>98%
C35H34N6O
10mg
Safety and Handling

Safety and Handling

Recommended Storage : Store at −20°C

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit