Learn More
CTAG1B Mouse anti-Human, Clone: OTI2A10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505948

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by CTAG1B is an antigen that is overexpressed in many cancers but that is also expressed in normal testis. This gene is found in a duplicated region of the X-chromosome and therefore has a neighboring gene of identical sequence.Specifications
CTAG1B | |
Monoclonal | |
Unconjugated | |
CTAG1B | |
CTAG1B | |
Full length human recombit protein of human CTAG1B produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2A10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
CT6.1, CTAG, CTAG1, ESO1, LAGE-2, LAGE2B, NY-ESO-1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1485 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.