Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTAG1B Mouse anti-Human, Clone: OTI4B6, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA505952

Catalog No. 50-167-6336


Add to cart

Description

Description

The protein encoded by CTAG1B is an antigen that is overexpressed in many cancers but that is also expressed in normal testis. This gene is found in a duplicated region of the X-chromosome and therefore has a neighboring gene of identical sequence.
Specifications

Specifications

CTAG1B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Liquid
CTAG1B
Full length human recombit protein of human CTAG1B produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
OTI4B6
Unconjugated
CTAG1B
CT6.1, CTAG, CTAG1, ESO1, LAGE-2, LAGE2B, NY-ESO-1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1485
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit