Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTAG1B Monoclonal Antibody (OTI9E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Supplier: OriGene TA505975
Description
The protein encoded by CTAG1B is an antigen that is overexpressed in many cancers but that is also expressed in normal testis. This gene is found in a duplicated region of the X-chromosome and therefore has a neighboring gene of identical sequence.Specifications
CTAG1B | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
CT6.1, CTAG, CTAG1, ESO1, LAGE-2, LAGE2B, NY-ESO-1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1485 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI9E7 | |
Unconjugated | |
CTAG1B | |
CTAG1B | |
Full length human recombit protein of human CTAG1B produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction