Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTAG1B Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI5H4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270486JF646

Catalog No. N270486J646


Add to cart

Description

Description

CTAG1B Monoclonal antibody specifically detects CTAG1B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

CTAG1B
OTI5H4
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
CTAG1B
Full length recombinant protein of human CTAG1B (NP_001318) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Autoimmunogenic cancer/testis antigen NY-ESO-1, cancer antigen 3, cancer/testis antigen 1, cancer/testis antigen 1B, Cancer/testis antigen 6.1, CT6.1L antigen family member 2, CTAG1CTAG1A, ESO1CTAG, LAGE2, LAGE2A, LAGE2BLAGE-2, New York esophageal squamous cell carcinoma 1, NY-ESO-1
Mouse
IgG1
0.1 mL
Primary
1485.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit