Learn More
Aviva Systems Biology Corp MIA2 Antibody - middle region (ARP47994_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47994_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTAGE5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CTAGE5. Alternate names: MEA6, MGEA, MGEA11, MGEA6ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-59696