Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTBP2 Antibody (OTI4D4), FITC, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270487F

Catalog No. NBP270487F


Add to Cart

Description

Description

CTBP2 Monoclonal specifically detects CTBP2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications

Specifications

CTBP2
Monoclonal
FITC
PBS with 0.05% Sodium Azide
CTBP2
Recombinant protein fragment corresponding to amino acids 233-336 of human CTBP2 (NP_073713) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Immunohistochemistry
OTI4D4
Immunohistochemistry
CtBP2, C-terminal binding protein 2, 10ribeye, C-terminal-binding protein 2
Mouse
Affinity Purified
Cell Cycle and Replication, Wnt Signaling Pathway
1488.0
Store at 4°C in the dark.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit