Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CtBP2 Monoclonal Antibody (OTI4D4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526926
Description
CtBP2 Monoclonal Antibody for Western Blot, IHC (P)
This gene produces alternative transcripts encoding two distinct proteins. One protein is a transcriptional repressor, while the other isoform is a major component of specialized synapses known as synaptic ribbons. Both proteins contain a NAD+ binding domain similar to NAD+-dependent 2-hydroxyacid dehydrogenases. A portion of the 3' untranslated region was used to map this gene to chromosome 21q21. 3; however, it was noted that similar loci elsewhere in the genome are likely. Blast analysis shows that this gene is present on chromosome 10.
Specifications
CtBP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P56545 | |
CTBP2 | |
Human recombinant protein fragment corresponding to amino acids 233-336 of CTBP2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4D4 | |
Unconjugated | |
CTBP2 | |
AA407280; CTBP2; C-terminal binding protein 2; C-terminal-binding protein 2; D7Ertd45e; gene trap ROSA b-geo 6; Gtrgeo6; Ribeye; RP11-59C5.2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1488 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction