Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTBP2 Antibody (OTI4D4), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270487AF700
Description
CTBP2 Monoclonal specifically detects CTBP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CTBP2 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CTBP2 | |
Recombinant protein fragment corresponding to amino acids 233-336 of human CTBP2 (NP_073713) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI4D4 | |
Western Blot, Immunohistochemistry | |
CtBP2, C-terminal binding protein 2, 10ribeye, C-terminal-binding protein 2 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Wnt Signaling Pathway | |
1488.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction