Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTBP2 Mouse anti-Human, HRP, Clone: OTI4D4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270487H

Catalog No. NBP270487H


Add to cart

Description

Description

CTBP2 Monoclonal antibody specifically detects CTBP2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

CTBP2
OTI4D4
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
CTBP2
Recombinant protein fragment corresponding to amino acids 233-336 of human CTBP2 (NP_073713) produced in E.coli.
Immunogen affinity purified
Cell Cycle and Replication, Wnt Signaling Pathway
Primary
1488.0
Immunohistochemistry, Western Blot
HRP
PBS with No Preservative
CtBP2, C-terminal binding protein 2, 10ribeye, C-terminal-binding protein 2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit