Learn More
Invitrogen™ CTCF Monoclonal Antibody (1D11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515792
Description
MA5-15792 targets CTCF in indirect ELISA, WB applications and shows reactivity with Human and Non-human primate samples. The MA5-15792 immunogen is purified recombinant fragment of human CTCF expressed in E. Coli. MA5-15792 detects CTCF which has a predicted molecular weight of approximately 83kDa.
Transcriptional factor CTCF is a ubiquitously expressed 11-zinc finger domain containing, DNA-binding nuclear phosphoprotein that is involved in multiple aspects of normal gene regulation including transcriptional repression and activation, gene silencing, chromatin insulation and regulation of imprinted sites. CTCF uses different combinations of ZF domains and interacts with the CCCTC motif on the DNA. It can bind to HAT- and HDAC-containing complexes thereby influencing transcriptional activation or repression respectively. It is a multifunctional protein involved in the transactivation of APPB gene, silencing of c-myc gene, and insulation of human beta-globin and DMI myotonic dystrophy locus and imprinting control of IGF2 and H19 regions. CTCF has also been shown to regulate the transcription of Pax6 and IRAK2 promoter thereby opening a therapeutic avenue. Mutations in this gene have been associated with invasive breast cancers, prostate cancers and Wilms' tumors.
Specifications
CTCF | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P49711, Q61164 | |
CTCF | |
Purified recombinant fragment of human CTCF expressed in E. Coli. | |
RUO | |
10664, 13018 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1D11 | |
Unconjugated | |
CTCF | |
11 zinc finger transcriptional repressor; 11-zinc finger protein; AW108038; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein); Ctcf; ctcf protein; CTCFL paralog; im:7152666; MRD21; OTTHUMP00000197610; Transcriptional repressor CTCF; Unknown (protein for IMAGE:8125283) | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse, Monkey | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.