Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTDSP2 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI7F5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271552AF700

Catalog No. N271552A700


Add to cart

Description

Description

CTDSP2 Monoclonal antibody specifically detects CTDSP2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

CTDSP2
OTI7F5
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
CTDSP2
Full length recombinant protein of human CTDSP2 (NP_005721) produced in E.coli.
Immunogen affinity purified
Protein Phosphatase
Primary
10106.0
Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 2, conserved gene amplified in osteosarcoma, CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) smallphosphatase 2, NIF2, Nuclear LIM interactor-interacting factor 2, OS4NLI-interacting factor 2, Protein OS-4, PSR2, SCP2EC 3.1.3.16, Small CTD phosphatase 2, Small C-terminal domain phosphatase 2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit