Learn More
CTDSP2 Mouse anti-Human, Clone: OTI7F5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808714
Catalog No. 50-166-9029
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CTDSP2 is one of several small CTD phosphatases (SCP). It has been proposed that SCP activity is an evolutionarily conserved transcriptional regulator that acts globally to silence neuronal genes. In addition, expression of phosphatase CTDSP2 mRNA isenhanced in several cancer cell lines with amplification of the CTDSP2 and CDK4 genes, and CTDSP2 coamplifies with CDK4 in some primary sarcomas. CTDSP2 may contribute to the development of a subset of sarcomas.
Specifications
Specifications
CTDSP2
Monoclonal
Unconjugated
CTDSP2
Lyophilized
CTDSP2
Full length human recombit protein of human CTDSP2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI7F5
PBS with 8% trehalose and no preservative; pH 7.3
O14595
OS4, PSR2, SCP2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10106