Learn More
CTDSPL Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB30013
Catalog No. 89-033-301
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human CTDSPL.
  • High specificity
  • High reactivity
Specifications
Specifications
CTDSPL
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
NM_005808
C3orf8|HYA22|PSR1|RBSP3|SCP3
Rabbit
Affinity purification
RUO
Primary
10217
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human CTDSPL.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
CTDSPL
A synthetic peptide corresponding to N-terminus of human CTDSPL.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)