Learn More
CTGF/CCN2 Mouse anti-Human, DyLight 405, Clone: MM0038-6B13, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11061004V
Catalog No. NB11061004V
Add to cart
Description
Description
CTGF/CCN2 Monoclonal specifically detects CTGF/CCN2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CTGF/CCN2
MM0038-6B13
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
CCN2
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with recombinant human Connective Tissue Growth Factor (CTGF).
Protein G purified
Angiogenesis, Cancer, Cell Biology, Cytokine Research, Growth and Development, Immunology, Innate Immunity
Primary
1490
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human