Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTGF Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 239361AP

Catalog No. 50-173-4169


Add to cart

Description

Description

Connective Tissue Growth Factor (CTGF), also called CCN2, is a member of the CCN family. CTGF is a secreted protein produced by umbilical veins and vascular endothelial cells. CTGF possesses an Insulin-like Growth Factor (IGF)-binding domain, a thrombospondin type 1 domain, and a cysteine knot region. CTGF plays important roles in the proliferation and differentiation of chondrocytes, induces angiogenesis, and promotes cell adhesion in fibroblasts, endothelial, and epithelial cells.
Specifications

Specifications

CTGF
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P29268, P29279
CCN family member 2, CCN2, CTGF, HCS24, IBP 8, IGF binding protein 8, IGFBP 8, IGFBP8, NOV2
Rabbit
IgG
150 μL
-20°C
Primary
14219, 1490
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CTGF
Liquid
Ctgf
CTGF Fusion Protein Ag21008
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit