Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTH Mouse anti-Canine, Human, Clone: OTI1E12, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502442

Catalog No. 50-167-3498


Add to cart

Description

Description

This gene encodes a cytoplasmic enzyme in the trans-sulfuration pathway that converts cystathione derived from methionine into cysteine. Glutathione synthesis in the liver is dependent upon the availability of cysteine. Mutations in this gene cause cystathioninuria. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
Specifications

Specifications

CTH
Monoclonal
0.32 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P32929
cystathionine gamma-lyase
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1491, 479991
Immunohistochemistry (Paraffin), Western Blot
OTI1E12
Unconjugated
CTH
Liquid
CTH
Full length human recombit protein of human CTH produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit