Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTHRC1 Rabbit anti-Human, Clone: 62, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | CTHRC1 |
---|---|
Clone | 62 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
CTHRC1 Monoclonal specifically detects CTHRC1 in Human samples. It is validated for ELISA.Specifications
CTHRC1 | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
115908 | |
IgG | |
Protein A purified |
62 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, Protein NMTC1 | |
Recombinant Human CTHRC1 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title