Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTHRC1 Antibody (166), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289947UV
Description
CTHRC1 Monoclonal specifically detects CTHRC1 in Human samples. It is validated for Western Blot.Specifications
CTHRC1 | |
Monoclonal | |
DyLight 350 | |
collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, Protein NMTC1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CTHRC1 (NP_612464.1; Met1-Lys243). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
166 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
115908 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction