Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTHRC1 Rabbit anti-Human, DyLight 405, Clone: 166, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289947V

Catalog No. NB110301


Description

Description

CTHRC1 Monoclonal specifically detects CTHRC1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CTHRC1
166
50 mM Sodium Borate
collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, Protein NMTC1
This antibody was obtained from a rabbit immunized with purified, recombinant Human CTHRC1 (NP_612464.1; Met1-Lys243).
Protein A purified
RUO
Primary
115908
Western Blot
DyLight 405
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit