Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTI Powder-Free Nitrile Exam Gloves

Manufacturer:  CTI NPX101

 View more versions of this product

Catalog No. 19-050-342

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications

Specifications

Safety
Tapered, beaded
Yes
Small
5 mil
White
25.4 cm
Textured fingertips
Ambidextrous
Nitrile
14
Powder-free
10 in.
ASTM D 3578-95, MIL STD-105E, 21 CFR 177-2600, FDA
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit