Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CTIF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553509
Description
Immunogen sequence: TFDSFSGATW DLQPEKLDFT QFHRKVRHTP KQPLPHIDRE GCGKGKLEDG DGINLNDIEK VLPAWQGYHP MPHEVEIAHT KKLFR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

Specifications
CTIF | |
Polyclonal | |
Unconjugated | |
CTIF | |
cap binding complex dependent translation initiation factor; CBP80/20-dependent translation initiation factor; Ctif; Gm672; KIAA0427 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9811 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43310 | |
CTIF | |
Recombinant protein corresponding to Human CTIF. Recombinant protein control fragment (Product #RP-92416). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction