Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTLA-4 Antibody (WKH203), Alexa Fluor™ 488

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063254AF488
Description
CTLA-4 Monoclonal specifically detects CTLA-4 in Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
CTLA-4 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CTLA4 | |
Purified rCTLA-4hIg fusion protein. | |
0.25 mL | |
Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
1493 | |
Rat | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
WKH203 | |
Western Blot, Flow Cytometry, ELISA | |
CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
NB100-63254 recognizes rat CD152, also known as cytotoxic-T-lymphocyte antigen-4 (CTLA-4), which is similar in structure to CD28 and also binds ligands CD80 and CD86. CD152 is expressed by activated T-lymphocytes. Studies suggest that CD152 is also expressed by regulatory T-lymphocytes. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction