Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTLA-4 (CD152) Chimeric Recombinant Rabbit Monoclonal Antibody (9D9), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA548129

Encompass

Catalog No. PIMA548129


Add to Cart

Description

Description

Specificity: The antibody is specific for CTLA-4.

CTLA-4 (CD152) Chimeric Recombinant Monoclonal antibody specifically detects CTLA-4 (CD152) Chimeric in Mouse samples. It is validated for Immunocytochemistry, Inhibition Assays
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CTLA-4 (CD152) Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
Q9D659
Rabbit
Affinity chromatography
RUO
74048
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Immunocytochemistry, Inhibition Assays
9D9
Unconjugated
VSIR
ALPS5; CD; CD152; CD152 antigen; CD152 isoform; CD152 protein; CD152 protein precursor; celiac disease 3; CELIAC3; Ctla4; CTLA-4; cytotoxic T lymphocyte associated antigen 4 short spliced form; cytotoxic T-lymphocyte associated protein 4; cytotoxic T-lymphocyte protein 4; cytotoxic T-lymphocyte protein 4 isoform CTLA4-TM; cytotoxic T-lymphocyte-associated antigen 4; cytotoxic T-lymphocyte-associated protein 4; cytotoxic T-lymphocyte-associated serine esterase-4; EGK_04712; GRD4; GSE; ICOS; IDDM12; insulin-dependent diabetes mellitus 12; ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4; Ly-56; sCTLA4; soluble form; transmembrane form
The antibody was generated by immunising a PD-1H-KO mouse with mouse PD-1H Ig fusion protein and adjuvant.
200 μg
Primary
Mouse
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit