Learn More
CTLA-4 Human anti-Human, Alexa Fluor 594, Clone: Hu111, R&D Systems
Human Monoclonal Antibody
Manufacturer:  R&D Systems FAB9928T100UG
Catalog No. FAB9928T100
Add to cart
This item is not returnable. View return policy
Description
Description
CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CTLA-4
Hu111
Flow Cytometry 0.25-1 μg/10^6 cells
Purified, Supernatant, Conjugated
Human
IgG1
100 μg
Primary
1493
Human
Flow Cytometry
Alexa Fluor 594
Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide.
CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4
Human CTLA-4
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects Human CTLA-4 based on Ipilimumab therapeutic antibody.This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Ipilimumab. This product is for research use only.