Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTLA-4 Human anti-Human, PE, Clone: Hu111, R&D Systems™
Human Monoclonal Antibody
Supplier: R&D Systems FAB9928P100
This item is not returnable.
View return policy
Description
CTLA-4 Monoclonal specifically detects CTLA-4 in Human samples. It is validated for Flow Cytometry.Specifications
CTLA-4 | |
Monoclonal | |
PE | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
Human | |
Protein A or G purified from cell culture supernatant | |
RUO | |
1493 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Flow Cytometry | |
Hu111 | |
Flow Cytometry 10 uL/10^6 cells | |
CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
Human CTLA-4 | |
100 Tests | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title