Learn More
CTLA-4 Rabbit anti-Human, Alexa Fluor 488, Clone: 2188B, R&D Systems
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB3253G100UG
Catalog No. FB3253G100
Add to cart
This item is not returnable. View return policy
Description
Description
CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CTLA-4
2188B
Flow Cytometry 0.25-1 μg/106 cells
P16410
CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4
Chinese hamster ovary cell line CHO-derived recombinant human CTLA-4, Ala37-Phe162, Accession No. P16410
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human CTLA-4 in direct ELISAs.
Flow Cytometry
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Rabbit
IgG
100 μg
Primary
1493.0
Human