Learn More
CTLA-4 Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0199-6G10, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212212AF532
Catalog No. NP212212A32
Add to cart
Description
Description
CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Mouse samples. It is validated for Western Blot,Flow Cytometry.
Specifications
Specifications
CTLA-4
RM0199-6G10
Western Blot, Flow Cytometry
CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Flow Cytometry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CTLA4
Mouse CTLA4
0.1 mL
Primary
1493