Learn More
CTLA4 Mouse anti-Human, Clone: OTI8D12, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF807714
Catalog No. 50-166-8434
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CTLA4 (Cytotoxic T-Lymphocyte Antigen 4) is a CD28-family receptor expressed on mainly CD4+ T cells. It binds the same ligands as CD28 (CD80 and CD86 on B cells and dendritic cells), but with higher affinity. However, in contrast to CD28 which enhances cell function when bound at the same time as the T cell receptor, CTLA4 inhibits the T cell and prevents it from functioning. Intracellular CTLA4 is also found in regulatory T cells and may be important to their function.
Specifications
Specifications
CTLA4
Monoclonal
Unconjugated
CTLA4
Lyophilized
CTLA4
Full length human recombit protein of human CTLA4 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI8D12
PBS with 8% trehalose and no preservative; pH 7.3
P16410
ALPS5, CD, CD152, CELIAC3, CTLA-4, GRD4, GSE, IDDM12
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1493