Learn More
Abnova™ CTNNA3 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00029119T01
Catalog No. 89-940-893
Add to cart
Specifications
Specifications
Western Blot
293T
BC065819.1
pCMV-CTNNA3 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
CTNNA3 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 56.87kDa
Transient overexpression cell lysate was tested with anti-CTNNA3 antibody (Mfr. No. H00029119-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)