Learn More
Aviva Systems Biology Corp CTNNB1 Antibody - C-terminal region (P100601_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100601_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTNNB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C-terminal region of human CTNNB1. Alternate names: CTNNB, DKFZp686D02253, FLJ25606, FLJ37923ApplicationCHIP, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Zebrafish
Catalog No. 50-014-62657