Learn More
CTNNB1 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: EC-3, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB19614
Catalog No. 89-336-139
Add to cart
Description
Description
CTNNB1 Monoclonal antibody specifically detects CTNNB1 in Human, Mouse, Rat samples. It is validated for IP, ICC, WB-Ce, IHC-P, IF.
Specifications
Specifications
CTNNB1
EC-3
catenin (cadherin-associated protein), beta 1, 88kDa
Liquid
Rabbit
IgG
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
CTNNB1
CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
A synthetic peptide corresponding to human CTNNB1
100 μg
Primary
1499