Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTNNBL1 Mouse anti-Human, Biotin, Clone: 1E4F5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237252B

Catalog No. NBP237252B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CTNNBL1
1E4F5
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
CTNNBL1
Purified recombinant fragment of human CTNNBL1 (AA: 390-557) expressed in E. Coli.
Protein A or G purified
Apoptosis, Cancer
Primary
56259
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
beta-catenin-like protein 1, C20orf33, catenin, beta like 1, dJ633O20.1, FLJ21108, NAPchromosome 20 open reading frame 33, nuclear associated protein, Nuclear-associated protein, NYD-SP19, P14, P14L, PP8304, Testis development protein NYD-SP19
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit