Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTNS Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549199

Catalog No. PA549199

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cystine/H(+) symporter thought to transport cystine out of lysosomes. Plays an important role in melanin synthesis, possibly by preventing melanosome acidification and subsequent degradation of tyrosinase TYR.
Specifications

Specifications

CTNS
Polyclonal
PBS with 0.09% sodium azide
O60931
Cystinosin, CTNS
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1497
Western Blot
Unconjugated
CTNS
Liquid
CTNS
KLH conjugated synthetic peptide between 341-367 amino acids from the C-terminal region of human CTNS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit