Learn More
Alfa Aesar™ CTOP
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66263MCR
 View more versions of this product
Catalog No. AAJ66263MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1062.27
1mg
D-Phe-Cys-Tyr-D-Trp-Orn-Thr-Pen-Thr-NH2 (Disulfide bridge: Cys2-Pen7)
-30° to -10°C
CTOP
HPLC
Solid
RUO
Cys2, Tyr3, Orn5, Pen7 amide