Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTPS Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550942

Catalog No. PA550942


Add to cart

Description

Description

CTPS Polyclonal Antibody detects endogenous levels of CTPS protein.

The catalytic conversion of UTP to CTP is accomplished by the enzyme cytidine-5-prime-triphosphate synthetase. The enzyme is important in the biosynthesis of phospholipids and nucleic acids, and plays a key role in cell growth, development, and tumorigenesis.The catalytic conversion of UTP to CTP is accomplished by the enzyme cytidine-5-prime-triphosphate synthetase. The enzyme is important in the biosynthesis of phospholipids and nucleic acids, and plays a key role in cell growth, development, and tumorigenesis. The region to which the CTPS gene has been mapped is the location of breakpoints involved in several tumor types.The catalytic conversion of UTP to CTP is accomplished by the enzyme cytidine-5-prime-triphosphate synthetase. The enzyme is important in the biosynthesis of phospholipids and nucleic acids, and plays a key role in cell growth, development, and tumorigenesis. The region to which the CTPS gene has been mapped is the location of breakpoints involved in several tumor types.
Specifications

Specifications

CTPS
Polyclonal
Unconjugated
CTPS1
Liquid
Ctps, Ctps1
Synthesized peptide derived from the N-terminal region of human CTPS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P17812, P70698
CTPS1, CTPS, CTP synthase 1, CTP synthetase 1, UTP--ammonia ligase 1
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1503, 313560, 51797
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit